missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-nitrobenzoic acid, Thermo Scientific™

Catalog Number p-75525
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
943-14-6
C7H4BrNO4
246.02
MFCD00134558
UVFWYVCDRKRAJH-UHFFFAOYSA-N
2-bromo-5-nitrobenzoic acid, 2-bromo-5-nitrobenzic acid, benzoic acid, 2-bromo-5-nitro, 2-bromo-5-nitrobenzoicacid, 2-bromo-5-nitro benzoic acid, pubchem3821, acmc-209rrc, ksc494m1r, paragos 440009, timtec-bb sbb003179
243025
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)Br
This item is not returnable. View return policy
943-14-6
C7H4BrNO4
246.02
MFCD00134558
UVFWYVCDRKRAJH-UHFFFAOYSA-N
2-bromo-5-nitrobenzoic acid, 2-bromo-5-nitrobenzic acid, benzoic acid, 2-bromo-5-nitro, 2-bromo-5-nitrobenzoicacid, 2-bromo-5-nitro benzoic acid, pubchem3821, acmc-209rrc, ksc494m1r, paragos 440009, timtec-bb sbb003179
243025
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)Br
Melting Point 180°C to 181°C
Color Beige to Brown
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 246.02
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material 2-Bromo-5-nitrobenzoic acid
compliance-icons
Product Identifier
  • 2-Bromo-5-nitrobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'