missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-nitroaniline, 98%, Thermo Scientific Chemicals

Catalog Number 10738932
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
10403-47-1
C6H5BrN2O2
217.02
MFCD00051578
BAAUCXCLMDAZEL-UHFFFAOYSA-N
benzenamine, 2-bromo-5-nitro, 2-bromo-5-nitrobenzenamine, 3-amino-4-bromonitrobenzene, 2-bromo-5-nitro-phenylamine, 2-bromo-5-nitrophenylamine, pubchem21903, 2-bromo 5-nitroaniline, 2-bromo-5-nitro-aniline, 2-bromo-5-nitro aniline, acmc-2098ai
82607
2-bromo-5-nitroaniline
C1=CC(=C(C=C1[N+](=O)[O-])N)Br
This item is not returnable. View return policy
10403-47-1
C6H5BrN2O2
217.02
MFCD00051578
BAAUCXCLMDAZEL-UHFFFAOYSA-N
benzenamine, 2-bromo-5-nitro, 2-bromo-5-nitrobenzenamine, 3-amino-4-bromonitrobenzene, 2-bromo-5-nitro-phenylamine, 2-bromo-5-nitrophenylamine, pubchem21903, 2-bromo 5-nitroaniline, 2-bromo-5-nitro-aniline, 2-bromo-5-nitro aniline, acmc-2098ai
82607
2-bromo-5-nitroaniline
C1=CC(=C(C=C1[N+](=O)[O-])N)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 138°C to 142°C
Color Khaki to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: slightly soluble in water
Formula Weight 217.02
Percent Purity 98%
Physical Form Crystalline Powder or Fine Crystalline Needles
Chemical Name or Material 2-Bromo-5-nitroaniline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-5-nitroaniline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'