missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-(methanesulfonyl)pyridine, 97%, ACROS Organics™

Brand:  Acros Organics 446620050

UNSPSC : 12161500

Code : 29

Additional Details : CAS Number : 343262-51-1 Weight : 0.00500kg


Quantity 5g

 View more versions of this product

Product Code. 11926481

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications Description & Specifications

Chemical Identifiers

2-Bromo-5-(methanesulfonyl)pyridine
97%
C6H6BrNO2S
236.08

Specifications

2-Bromo-5-(methanesulfonyl)pyridine
5g
C6H6BrNO2S
343262-51-1
97%
236.08

Safety and Handling

Product Identifier
  • 2-Bromo-5-(methanesulfonyl)pyridine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Documentation

Technical Documents