missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-iodobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog Number 11314126
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
25252-00-0
C7H4BrIO2
326.915
MFCD00079716
QPKKBDSNZFSSOD-UHFFFAOYSA-N
2-bromo-5-iodobenzoicacid, 2-bromo-5-iodo-benzoic acid, benzoic acid, 2-bromo-5-iodo, pubchem3787, acmc-209gii, intermediates-zcf02680, ksc497i9t, zerenex e/9071914, 2-bromo-5-iodobenzoic acid
2735572
2-bromo-5-iodobenzoic acid
C1=CC(=C(C=C1I)C(=O)O)Br
This item is not returnable. View return policy
25252-00-0
C7H4BrIO2
326.915
MFCD00079716
QPKKBDSNZFSSOD-UHFFFAOYSA-N
2-bromo-5-iodobenzoicacid, 2-bromo-5-iodo-benzoic acid, benzoic acid, 2-bromo-5-iodo, pubchem3787, acmc-209gii, intermediates-zcf02680, ksc497i9t, zerenex e/9071914, 2-bromo-5-iodobenzoic acid
2735572
2-bromo-5-iodobenzoic acid
C1=CC(=C(C=C1I)C(=O)O)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 174°C to 176°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 326.92
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-iodobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'