missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-hydroxypyrazine, 97%, ACROS Organics™

Brand:  Acros Organics 435890010

Code : 29

Additional Details : CAS Number : 18437-66-6 Weight : 0.05100kg


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10586459

  • £184.90 / 1g


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

2-Bromo-5-hydroxypyrazine
96.0
≥96.0%
C4H3BrN2O
MFCD06245330
374063-92-0
100.0
97%
174.98

Specifications

2-Bromo-5-hydroxypyrazine
96.0
Authentic
White to light brown
≥96.0%
1g
174.98
374063-92-0
100.0
Glass bottle
Crystalline Powder
97%
C4H3BrN2O
MFCD06245330
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Bromo-5-hydroxypyrazine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS