missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromo-5-hydroxy-3-methylpyridine, 97%

Brand:  Alfa Aesar™ H64567.03

Code : NEW

Additional Details : CAS Number : 1003711-43-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15411849

  • £168.60 / 1g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

2-Bromo-5-hydroxy-3-methylpyridine
188.02
2-Bromo-5-hydroxy-3-picoline
1003711-43-0
MFCD09839267

Specifications

2-Bromo-5-hydroxy-3-methylpyridine
White
97%
188.02
2-Bromo-5-hydroxy-3-picoline
1003711-43-0
Powder
1g
MFCD09839267
Videos
SDS