missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluoropyridine, 98%, ACROS Organics™

Brand:  Acros Organics 397970010

Code : 29

Additional Details : CAS Number : 41404-58-4 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 1 Pack group : III


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10557434

  • £26.70 / 1g


Estimated Shipment
15-05-2018
Add to basket


Specifications

Chemical Identifiers

2-Bromo-5-fluoropyridine
97.5
98%
C5H3BrFN
MFCD00234011
41404-58-4
100.0
98%
175.98

Specifications

2-Bromo-5-fluoropyridine
97.5
Authentic
1.5390 to 1.5410 (20°C, 589nm)
30.0° to 31.0°C
98%
98%
73°C
175.98
41404-58-4
100.0
Glass bottle
Colorless to slightly yellow
Liquid
83.0°C (44.0 mmHg)
1g
C5H3BrFN
MFCD00234011
Videos
SDS