missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-5-fluorobenzyl alcohol, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
2-Bromo-5-fluorobenzyl alcohol is used as intermediates in pharmaceutical and organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-5-fluorobenzyl alcohol is used as intermediates in pharmaceutical and organic synthesis.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store in a cool, dark and well-ventilated place. Store away from incompatible materials such as oxidizing agents.
2-Bromo-5-fluorobenzyl alcohol is used as intermediates in pharmaceutical and organic synthesis.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store in a cool, dark and well-ventilated place. Store away from incompatible materials such as oxidizing agents.
Specifications
Melting Point | 91°C to 94°C |
Color | Yellow |
Quantity | 1 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 205.02 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | 2-Bromo-5-fluorobenzyl alcohol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'