missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluorobenzotrifluoride, 97%, ACROS Organics™

Brand:  Acros Organics 442570050

Code : 29

Additional Details : CAS Number : 393-37-3 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 11914551

  • £72.60 / 5g


Estimated Shipment
09-02-2018
Add to basket


Specifications

Chemical Identifiers

2-Bromo-5-fluorobenzotrifluoride
96.0
≥96.0%
C7H3BrF4
393-37-3
100.0
97%
243

Specifications

2-Bromo-5-fluorobenzotrifluoride
96.0
Authentic
1.75g/mL
Liquid
97%
C7H3BrF4
393-37-3
100.0
Glass bottle
Clear colorless to pale yellow
≥96.0%
5g
243
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Bromo-5-fluorobenzotrifluoride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/A§/if you feel unwell.

SDS