missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-chloro-3-nitropyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-7055475
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
75806-86-9
C5H2BrClN2O2
237.437
MFCD07368982
VFMIDVIAQMFGGP-UHFFFAOYSA-N
2-bromo-3-nitro-5-chloropyridine, pyridine, 2-bromo-5-chloro-3-nitro, 2-bromo-5-chloro-3-nitro-pyridine, pubchem5951, acmc-1bggp, ksc495s5l, 2-bromo-5-chloro-3-nitro pyridine, 3-nitro-2-bromo-5-chloropyridine, 5-chloro-2-bromo-3-nitropyridine
280681
2-bromo-5-chloro-3-nitropyridine
C1=C(C(=NC=C1Cl)Br)[N+](=O)[O-]
This item is not returnable. View return policy
75806-86-9
C5H2BrClN2O2
237.437
MFCD07368982
VFMIDVIAQMFGGP-UHFFFAOYSA-N
2-bromo-3-nitro-5-chloropyridine, pyridine, 2-bromo-5-chloro-3-nitro, 2-bromo-5-chloro-3-nitro-pyridine, pubchem5951, acmc-1bggp, ksc495s5l, 2-bromo-5-chloro-3-nitro pyridine, 3-nitro-2-bromo-5-chloropyridine, 5-chloro-2-bromo-3-nitropyridine
280681
2-bromo-5-chloro-3-nitropyridine
C1=C(C(=NC=C1Cl)Br)[N+](=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 175°C to 179°C
Color White
Quantity 1 g
UN Number UN2811
Beilstein 147540
Formula Weight 237.44
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-5-chloro-3-nitropyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'