missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-chloro-3-methylbenzo[b]thiophene, 97%, Maybridge™

Brand:  Maybridge KM08172DA

Code : 31

Additional Details : CAS Number : 175203-60-8 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Packaging Amber Glass Bottle
Quantity 1g

 View more versions of this product

Product Code. 10069381

  • £11.00 / 1g

Estimated Shipment: 01-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

175203-60-8
261.561
RWFPSUKDYYBMHU-UHFFFAOYSA-N
2735548
CC1=C(SC2=C1C=C(C=C2)Cl)Br
C9H6BrClS
MFCD00126363
2-bromo-5-chloro-3-methylbenzo b thiophene, benzo b thiophene, 2-bromo-5-chloro-3-methyl, maybridge1_004921, acmc-1bx9l, 2-bromo-5-chloro-3-methylbenzthiophene, 2-bromo-5-chloro-3-methylbenzothiophene, 2-bromanyl-5-chloranyl-3-methyl-1-benzothiophene
2-bromo-5-chloro-3-methyl-1-benzothiophene

Specifications

2-Bromo-5-chloro-3-methylbenzo[b]thiophene
1g
97.0
C9H6BrClS
2-bromo-5-chloro-3-methylbenzo b thiophene, benzo b thiophene, 2-bromo-5-chloro-3-methyl, maybridge1_004921, acmc-1bx9l, 2-bromo-5-chloro-3-methylbenzthiophene, 2-bromo-5-chloro-3-methylbenzothiophene, 2-bromanyl-5-chloranyl-3-methyl-1-benzothiophene
CC1=C(SC2=C1C=C(C=C2)Cl)Br
261.561
261.57
Amber Glass Bottle
175203-60-8
97%
MFCD00126363
RWFPSUKDYYBMHU-UHFFFAOYSA-N
2-bromo-5-chloro-3-methyl-1-benzothiophene
2735548
97%
Videos
SDS Documents
Certificates