missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number 15487908
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
43192-31-0
C8H7BrO2
215.05
MFCD09788636
ODISAUHBLBVQKC-UHFFFAOYSA-N
benzaldehyde, 2-bromo-4-methoxy, benzaldehyde,2-bromo-4-methoxy, 3-bromo-4-formylanisole, 2-bromo-p-anisaldehyde, pubchem12556, acmc-209jtd, ksc232o5d, 2-bromanyl-4-methoxy-benzaldehyde
11127641
2-bromo-4-methoxybenzaldehyde
COC1=CC(Br)=C(C=O)C=C1
This item is not returnable. View return policy
43192-31-0
C8H7BrO2
215.05
MFCD09788636
ODISAUHBLBVQKC-UHFFFAOYSA-N
benzaldehyde, 2-bromo-4-methoxy, benzaldehyde,2-bromo-4-methoxy, 3-bromo-4-formylanisole, 2-bromo-p-anisaldehyde, pubchem12556, acmc-209jtd, ksc232o5d, 2-bromanyl-4-methoxy-benzaldehyde
11127641
2-bromo-4-methoxybenzaldehyde
COC1=CC(Br)=C(C=O)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 215.04
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-4-methoxybenzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'