missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromo-4-fluoropyridine, 97%

Brand:  Alfa Aesar™ H64785.03

Code : NEW

Additional Details : CAS Number : 357927-50-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15462139

  • £30.60 / 1g


Estimated Shipment date
from Supplier 22-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Bromo-4-fluoropyridine
175.99
14158224
357927-50-5
MFCD09260909

Specifications

2-Bromo-4-fluoropyridine
1.5370
Liquid
1g
MFCD09260909
357927-50-5
1.699g/mL
97%
175.99
14158224
Videos
SDS