missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluorobenzylamine hydrochloride, 96%, Thermo Scientific Chemicals

Catalog Number 15427156
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
289038-14-8
C7H8BrClFN
240.5
MFCD00672922
PBCSNHSXUSMNOV-UHFFFAOYSA-N
2-bromo-4-fluorobenzylamine hydrochloride, 2-bromo-4-fluorophenyl methanamine hydrochloride, 1-2-bromo-4-fluorophenyl methanamine hydrochloride, 2-bromanyl-4-fluoranyl-phenyl methanamine hydrochloride
2773353
(2-bromo-4-fluorophenyl)methanamine;hydrochloride
C1=CC(=C(C=C1F)Br)CN.Cl
This item is not returnable. View return policy
289038-14-8
C7H8BrClFN
240.5
MFCD00672922
PBCSNHSXUSMNOV-UHFFFAOYSA-N
2-bromo-4-fluorobenzylamine hydrochloride, 2-bromo-4-fluorophenyl methanamine hydrochloride, 1-2-bromo-4-fluorophenyl methanamine hydrochloride, 2-bromanyl-4-fluoranyl-phenyl methanamine hydrochloride
2773353
(2-bromo-4-fluorophenyl)methanamine;hydrochloride
C1=CC(=C(C=C1F)Br)CN.Cl

2-Bromo-4-fluorobenzylamine hydrochloride used as a intermediate in pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4-fluorobenzylamine hydrochloride used as a intermediate in pharmaceutical.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 240.5
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Bromo-4-fluorobenzylamine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'