missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluoroanisole, 98%, Thermo Scientific Chemicals

Catalog Number p-7081073
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
452-08-4
C7H6BrFO
205.03
MFCD00012014
JIQXVIJARQLCOY-UHFFFAOYSA-N
2-bromo-4-fluoroanisole, benzene, 2-bromo-4-fluoro-1-methoxy, 2-bromo-4-fluoro anisole, 1-bromo-3-fluoro-6-methoxybenzene, 2-bromo-4-fluoro-methoxybenzene, 2-bromo-4-fluoro-1-methoxy-benzene, 1-bromo-5-fluoro-2-methoxybenzene, attercop-chm at106665, pubchem1972, 2-bromo-4-fluoroanisol
136292
2-bromo-4-fluoro-1-methoxybenzene
COC1=CC=C(F)C=C1Br
This item is not returnable. View return policy
452-08-4
C7H6BrFO
205.03
MFCD00012014
JIQXVIJARQLCOY-UHFFFAOYSA-N
2-bromo-4-fluoroanisole, benzene, 2-bromo-4-fluoro-1-methoxy, 2-bromo-4-fluoro anisole, 1-bromo-3-fluoro-6-methoxybenzene, 2-bromo-4-fluoro-methoxybenzene, 2-bromo-4-fluoro-1-methoxy-benzene, 1-bromo-5-fluoro-2-methoxybenzene, attercop-chm at106665, pubchem1972, 2-bromo-4-fluoroanisol
136292
2-bromo-4-fluoro-1-methoxybenzene
COC1=CC=C(F)C=C1Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.581
Boiling Point 88°C to 89°C (1 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.545
Quantity 10 g
Beilstein 2358880
Formula Weight 205.03
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-fluoroanisole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'