missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluoro-6-nitroaniline, tech. 90%, Thermo Scientific™

Catalog Number p-7054003
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
10472-88-5
C6H4BrFN2O2
235.01
MFCD07779528
HCYDUPDSEDHSQB-UHFFFAOYSA-N
2-bromo-4-fluoro-6-nitrobenzenamine, 2-bromo-4-fluoro-6-nitrophenylamine, benzenamine, 2-bromo-4-fluoro-6-nitro, 2-amino-3-bromo-5-fluoronitrobenzene, pubchem16316, ksc495m4t, 4-fluoro-2-bromo-6-nitroaniline, 4-fluoro-2-nitro-6-bromoaniline, 6-bromo-4-fluoro-2-nitroaniline, 2-bromo-4-fluoro-6-nitroaniline
292004
2-bromo-4-fluoro-6-nitroaniline
NC1=C(Br)C=C(F)C=C1[N+]([O-])=O
This item is not returnable. View return policy
10472-88-5
C6H4BrFN2O2
235.01
MFCD07779528
HCYDUPDSEDHSQB-UHFFFAOYSA-N
2-bromo-4-fluoro-6-nitrobenzenamine, 2-bromo-4-fluoro-6-nitrophenylamine, benzenamine, 2-bromo-4-fluoro-6-nitro, 2-amino-3-bromo-5-fluoronitrobenzene, pubchem16316, ksc495m4t, 4-fluoro-2-bromo-6-nitroaniline, 4-fluoro-2-nitro-6-bromoaniline, 6-bromo-4-fluoro-2-nitroaniline, 2-bromo-4-fluoro-6-nitroaniline
292004
2-bromo-4-fluoro-6-nitroaniline
NC1=C(Br)C=C(F)C=C1[N+]([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Bromo-4-fluoro-6-nitroaniline is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Color Orange
Quantity 1 g
UN Number UN2811
Formula Weight 235.01
Percent Purity 90%
Grade Technical
Physical Form Powder
Chemical Name or Material 2-Bromo-4-fluoro-6-nitroaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'