missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-chloroanisole, 98%, ACROS Organics™

Brand:  Acros Organics 366560250

Code : 29

Additional Details : CAS Number : 60633-25-2 Weight : 0.09063kg


Quantity 25mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10762833

  • £37.40 / 25mL


EU Stock 5
Add to basket


Specifications

Chemical Identifiers

2-Bromo-4-chloroanisole
60633-25-2
100.0
98%
BrC6H3(Cl)OCH3
MFCD00079705
98%
97.5
98%
C7H6BrClO
221.48

Specifications

2-Bromo-4-chloroanisole
60633-25-2
97.5
Glass bottle
1.6250g/mL
25mL
98%
C7H6BrClO
221.48
98%
Authentic
100.0
1.5850 to 1.5870 (20°C, 589nm)
Clear colorless to pale yellow
Liquid
98%
BrC6H3(Cl)OCH3
MFCD00079705
Videos
SDS