missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-chloroaniline 98%, ACROS Organics™

Brand:  Acros Organics 393470250

Code : 29

Additional Details : CAS Number : 873-38-1 Weight : 0.07500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10454515

  • £87.40 / 25g


Estimated Shipment
18-06-2018
Add to basket


Specifications

Chemical Identifiers

2-Bromo-4-chloroaniline
97.5
≥97.5%
C6H5BrClN
206.47
873-38-1
100.0
98%
ClC6H3(Br)NH2
MFCD00041313

Specifications

2-Bromo-4-chloroaniline
Authentic
100.0
Pale brown
25g
≥97.5%
C6H5BrClN
206.47
873-38-1
97.5
Glass bottle
62° to 66°C
Crystalline Powder
98%
ClC6H3(Br)NH2
MFCD00041313
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Bromo-4-chloroaniline

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS