missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,6-dichloroaniline, 98%, ACROS Organics™

Brand:  Acros Organics 399600250

Code : 29

Additional Details : CAS Number : 697-86-9 Weight : 0.07500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III


Packaging Glass bottle
Quantity 25g

 View more versions of this product

Product Code. 10766085

  • £21.10 / 25g


EU Stock 3
Add to basket


Specifications

Chemical Identifiers

2-Bromo-4, 6-dichloroaniline
97.5
98%
Cl2C6H2(Br)NH2
MFCD00040936
697-86-9
100.0
C6H4BrCl2N
240.91

Specifications

2-Bromo-4, 6-dichloroaniline
97.5
Glass bottle
98%
Cl2C6H2(Br)NH2
MFCD00040936
25g
697-86-9
100.0
98%
C6H4BrCl2N
240.91
80.0° to 81.0°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Bromo-4,6-dichloroaniline

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS