missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,5-dimethoxybenzoic acid, 97%, Thermo Scientific™

Catalog Number p-3757034
Encompass
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6286-46-0
C9H9BrO4
261.07
MFCD00017542
HWFCHCRFQWEFMU-UHFFFAOYSA-N
6-bromoveratric acid, benzoic acid, 2-bromo-4,5-dimethoxy, 2-bromo-4,5-dimethoxybenzoicacid, 6-bromo-3,4-dimethoxybenzoic acid, 6-bromveratrumsaure, acmc-1b7v8, ksc496e4l, rarechem al be 0015, timtec-bb sbb037916
222963
2-bromo-4,5-dimethoxybenzoic acid
COC1=CC(Br)=C(C=C1OC)C(O)=O
This item is not returnable. View return policy
6286-46-0
C9H9BrO4
261.07
MFCD00017542
HWFCHCRFQWEFMU-UHFFFAOYSA-N
6-bromoveratric acid, benzoic acid, 2-bromo-4,5-dimethoxy, 2-bromo-4,5-dimethoxybenzoicacid, 6-bromo-3,4-dimethoxybenzoic acid, 6-bromveratrumsaure, acmc-1b7v8, ksc496e4l, rarechem al be 0015, timtec-bb sbb037916
222963
2-bromo-4,5-dimethoxybenzoic acid
COC1=CC(Br)=C(C=C1OC)C(O)=O
CAS Min % 96.0
CAS Max % 100.0
Melting Point 188°C to 190°C
Color Cream to White or Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H2(Br)CO2H
Quantity 1 g
Formula Weight 261.07
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Bromo-4, 5-dimethoxybenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-4,5-dimethoxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'