missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,5-difluorotoluene, 98%, Thermo Scientific™

Catalog Number p-7028737
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
875664-38-3
C7H5BrF2
207.018
MFCD07777160
FKEURBCLFHOBDM-UHFFFAOYSA-N
2-bromo-4,5-difluorotoluene, 1-bromo-4,5-difluoro-2-methyl-benzene, 4,5-difluoro-2-methylbromobenzene, ksc495c2l, bromo-4,5-difluoro-2-methyl-benzene, 4,5-difluoro-2-methyl-1-bromobenzene, 1-brommo-4,5-difluoro-2-methyl-benzene, benzene,1-bromo-4,5-difluoro-2-methyl
44890785
1-bromo-4,5-difluoro-2-methylbenzene
CC1=CC(=C(C=C1Br)F)F
This item is not returnable. View return policy
875664-38-3
C7H5BrF2
207.018
MFCD07777160
FKEURBCLFHOBDM-UHFFFAOYSA-N
2-bromo-4,5-difluorotoluene, 1-bromo-4,5-difluoro-2-methyl-benzene, 4,5-difluoro-2-methylbromobenzene, ksc495c2l, bromo-4,5-difluoro-2-methyl-benzene, 4,5-difluoro-2-methyl-1-bromobenzene, 1-brommo-4,5-difluoro-2-methyl-benzene, benzene,1-bromo-4,5-difluoro-2-methyl
44890785
1-bromo-4,5-difluoro-2-methylbenzene
CC1=CC(=C(C=C1Br)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Refractive Index 1.508
Quantity 250 mg
Formula Weight 207.02
Percent Purity 98%
Chemical Name or Material 2-Bromo-4,5-difluorotoluene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'