missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromo-3-(trifluoromethyl)benzoic acid, 98%

Brand:  Alfa Aesar™ H63761.MD

Code : NEW

Additional Details : CAS Number : 177420-63-2 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15470619

  • £14.10 / 250mg


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Description

Description

Applications
2-Bromo-3-(trifluoromethyl)benzoic acid is used as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

2-Bromo-3-(trifluoromethyl)benzoic acid
177420-63-2
269.02
MFCD08532493

Specifications

2-Bromo-3-(trifluoromethyl)benzoic acid
Solid
269.02
250mg
177420-63-2
98%
MFCD08532493
Videos
SDS