missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-3-pyridinemethanol, 96%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-Bromo-3-bromomethylpyridine was obtained by reaction of the 2-bromo-3-hydroxymethylpyridine with PBr3 in dichloromethane. Reactant in the synthesis of 3-[2-bromophenyl(or pyridyl)methoxymethyl]benzo[4,5]imidazo[2,1-b]thiazoles.
Solubility
Soluble in methanol(almost transparency).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
2-Bromo-3-bromomethylpyridine was obtained by reaction of the 2-bromo-3-hydroxymethylpyridine with PBr3 in dichloromethane. Reactant in the synthesis of 3-[2-bromophenyl(or pyridyl)methoxymethyl]benzo[4,5]imidazo[2,1-b]thiazoles.
Solubility
Soluble in methanol(almost transparency).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Quantity | 250 mg |
Solubility Information | Soluble in methanol(almost transparency). |
Formula Weight | 188.02 |
Percent Purity | 96% |
Chemical Name or Material | 2-Bromo-3-pyridinemethanol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'