missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-methylbenzoic acid, 97%, ACROS Organics™

£46.90 - £161.10

Chemical Identifiers

CAS 53663-39-1
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.04
MDL Number MFCD00079721
InChI Key LSRTWJCYIWGKCQ-UHFFFAOYSA-N
Synonym 2-bromo-3-methyl-benzoic acid, 2-bromo-m-toluic acid, 2-bromo-3-carboxytoluene, 2-bromo-3-methylbenzoicacid, 3-methyl-2-bromobenzoic acid, benzoic acid, bromo-3-methyl, 2-brom-3-methylbenzoic acid, pubchem3805, acmc-1ay3t, ksc497o4r
PubChem CID 2735588
IUPAC Name 2-bromo-3-methylbenzoic acid
SMILES CC1=CC=CC(=C1Br)C(=O)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10205154 SDS View Documents Acros Organics
398860010
1g Glass bottle £46.90
1g View alternative products
Save Up to 
»
EU Stock 4 Please to purchase this item. Need a web account? Register with us today!
10051264 SDS View Documents Acros Organics
398860050
5g Glass bottle £161.10
5g View alternative products
Save Up to 
»
EU Stock 1 Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

53663-39-1
215.04
LSRTWJCYIWGKCQ-UHFFFAOYSA-N
2735588
CC1=CC=CC(=C1Br)C(=O)O
C8H7BrO2
MFCD00079721
2-bromo-3-methyl-benzoic acid, 2-bromo-m-toluic acid, 2-bromo-3-carboxytoluene, 2-bromo-3-methylbenzoicacid, 3-methyl-2-bromobenzoic acid, benzoic acid, bromo-3-methyl, 2-brom-3-methylbenzoic acid, pubchem3805, acmc-1ay3t, ksc497o4r
2-bromo-3-methylbenzoic acid

Specifications

2-Bromo-3-methylbenzoic acid
Glass bottle
135.0°C to 138.0°C
53663-39-1
100.0
C8H7BrO2
MFCD00079721
LSRTWJCYIWGKCQ-UHFFFAOYSA-N
2-bromo-3-methylbenzoic acid
2735588
Powder
Authentic
White
1g
96.0
96% min. (HPLC)
BrC6H3(CH3)COOH
2-bromo-3-methyl-benzoic acid, 2-bromo-m-toluic acid, 2-bromo-3-carboxytoluene, 2-bromo-3-methylbenzoicacid, 3-methyl-2-bromobenzoic acid, benzoic acid, bromo-3-methyl, 2-brom-3-methylbenzoic acid, pubchem3805, acmc-1ay3t, ksc497o4r
CC1=CC=CC(=C1Br)C(=O)O
215.04
215.04
97%