missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-3-methylbenzoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 135.0°C to 138.0°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (HPLC) |
Packaging | Glass bottle |
Linear Formula | BrC6H3(CH3)COOH |
Quantity | 1 g |
Formula Weight | 215.04 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'