missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromo-3-methylaniline, ≥97%

Brand:  Alfa Aesar™ H64014.03

Code : NEW

Additional Details : CAS Number : 54879-20-8 Weight : 0.00100kg Transport : UN number : 2810 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15441029

  • £79.80 / 1g


Estimated Shipment date
from Supplier 21-05-2018
Add to basket

Specifications

Chemical Identifiers

2-Bromo-3-methylaniline
186.05
UN2810
54879-20-8
MFCD06411367
2079474

Specifications

2-Bromo-3-methylaniline
1.6050
48°C (0.03mmHg)
Pale brown
≥97%
MFCD06411367
2079474
54879-20-8
1.489g/mL
1g
Liquid
186.05
UN2810
Videos
SDS