2-Bromo-3-methyl-3H-imidazole-4-carboxylic acid methyl ester, 95%, Maybridge


Overview
Brand: Maybridge

Manufacturer Part Number: MO07325DA

UNSPSC: 12350000

Code: 31

Additional Details:
Additional Details: CAS Number: 120781-02-4 Weight: 0.05100kg Transport: UN number : 2811 Chem class : 1 Pack group : III


Packaging Amber Glass Bottle
Quantity 1g

Product Code. 10113094

Quantity Price
1 £ 185.0 / 1g
Estimated Shipment date
from Supplier 03-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2-Bromo-3-methyl-3H-imidazole-4-carboxylic acid methyl ester
CAS 120781-02-4
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C6H7BrN2O2
Formula Weight 219.04
MDL Number MFCD02179563

Specification

Formula Weight 219.04
Quantity 1g
CAS Min % 95.0
Assay 95%
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 120781-02-4
Assay Percent Range 95%
MDL Number MFCD02179563
Packaging Amber Glass Bottle
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C6H7BrN2O2
Health Hazard 1 GHS Signal Word: Warning
Chemical Name or Material 2-Bromo-3-methyl-3H-imidazole-4-carboxylic acid methyl ester
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Warning

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.