missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-3-hydroxypyridine, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
2-Bromo-3-hydroxypyridine is used to produce 2-bromo-4,6-diiodo-3-pyridinol. It is also used in azo dyes and in the synthesis of pterocellin A by reacting with kojic acid and in synthesis of orelline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-3-hydroxypyridine is used to produce 2-bromo-4,6-diiodo-3-pyridinol. It is also used in azo dyes and in the synthesis of pterocellin A by reacting with kojic acid and in synthesis of orelline.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents and strong acids.
2-Bromo-3-hydroxypyridine is used to produce 2-bromo-4,6-diiodo-3-pyridinol. It is also used in azo dyes and in the synthesis of pterocellin A by reacting with kojic acid and in synthesis of orelline.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications
Melting Point | 182°C to 186°C |
Quantity | 5 g |
Beilstein | 109829 |
Solubility Information | Soluble in alcohol. Insoluble in water. |
Formula Weight | 174 |
Percent Purity | 99% |
Chemical Name or Material | 2-Bromo-3-hydroxypyridine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'