missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-3-furoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 2-bromo-3-furoic acid, 2-bromo-3-carboxyfuran, 2-bromo3-furancarboxylic acid, 2-bromo-3-furancarboxylic acid, 3-furancarboxylic acid,2-bromo, 2-bromanylfuran-3-carboxylic acid, 3-furancarboxylic acid, 2-bromo, 2-bromofuran-3-carboxylic acid |
Formula Weight | 190.98 |
Percent Purity | 97% |
Chemical Name or Material | 2-Bromo-3-furoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'