missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-fluoropyridine-4-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7055654
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1211530-89-0
C6H3BrFNO2
220.00
MFCD13185796
DHFIVEUMGCJILQ-UHFFFAOYSA-N
2-bromo-3-fluoroisonicotinic acid, 2-bromo-3-fluoroisonicotinicacid, 2-bromo-3-fluoro-4-picolic acid, 2-bromo-3-fluoro-4-pyridinecarboxylic acid, 2-bromo-3-fluoropyridin-4-carboxylicacid, 2-bromo-3-fluoropyridin-4-carboxylic acid
70680697
2-bromo-3-fluoropyridine-4-carboxylic acid
OC(=O)C1=C(F)C(Br)=NC=C1
This item is not returnable. View return policy
1211530-89-0
C6H3BrFNO2
220.00
MFCD13185796
DHFIVEUMGCJILQ-UHFFFAOYSA-N
2-bromo-3-fluoroisonicotinic acid, 2-bromo-3-fluoroisonicotinicacid, 2-bromo-3-fluoro-4-picolic acid, 2-bromo-3-fluoro-4-pyridinecarboxylic acid, 2-bromo-3-fluoropyridin-4-carboxylicacid, 2-bromo-3-fluoropyridin-4-carboxylic acid
70680697
2-bromo-3-fluoropyridine-4-carboxylic acid
OC(=O)C1=C(F)C(Br)=NC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 220
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-3-fluoropyridine-4-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'