missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromo-3-fluoro-6-methylpyridine, 98%

Brand:  Alfa Aesar™ H64900.03

Code : NEW

Additional Details : CAS Number : 374633-36-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15412309

  • £10.40 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Bromo-3-fluoro-6-methylpyridine
190.02
18506793
374633-36-0
MFCD03095061
2-Bromo-3-fluoro-6-picoline

Specifications

2-Bromo-3-fluoro-6-methylpyridine
Solid
1g
MFCD03095061
2-Bromo-3-fluoro-6-picoline
374633-36-0
98%
190.02
18506793
Videos
SDS