missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromo-3-fluoro-4-methylpyridine, 95%

Brand:  Alfa Aesar™ H64070.06

UNSPSC : 12352101

Code : NEW

Additional Details : CAS Number : 884494-37-5 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15431109

  • £119.78 / 5g


Estimated Shipment date
from Supplier 08-09-2017
Add to basket

Specifications

Chemical Identifiers

2-Bromo-3-fluoro-4-methylpyridine
190.02
19198641
884494-37-5
MFCD04112499
2-Bromo-3-fluoro-4-picoline

Specifications

2-Bromo-3-fluoro-4-methylpyridine
Colorless to pale yellow
95%
190.02
19198641
884494-37-5
Liquid
5g
MFCD04112499
2-Bromo-3-fluoro-4-picoline
Documentation

Technical Documents