missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromo-3-chlorobenzotrifluoride, 98%

Brand:  Alfa Aesar™ H64302.06

Code : NEW

Additional Details : CAS Number : 384-16-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15411449

  • £162.50 / 5g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Bromo-3-chlorobenzotrifluoride
259.46
21501675
384-16-7
MFCD00673988
2-Bromo-1-chloro-3-(trifluoromethyl)benzene

Specifications

2-Bromo-3-chlorobenzotrifluoride
5g
98%
MFCD00673988
2-Bromo-1-chloro-3-(trifluoromethyl)benzene
384-16-7
Solid
259.46
21501675
Videos
SDS