missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3,5-dinitrobenzoic acid, 98%, Thermo Scientific™

Catalog Number 15472059
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
116529-60-3
C7H3BrN2O6
291.013
MFCD00458630
BBKJKUFARRJPRP-UHFFFAOYSA-N
2-bromo-3,5-dinitrobenzoic acid, acmc-1bobs, 2-bromo-3,5-dinitrobenzoicacid, benzoic acid,2-bromo-3,5-dinitro
3769937
C1=C(C=C(C(=C1[N+](=O)[O-])Br)C(=O)O)[N+](=O)[O-]
This item is not returnable. View return policy
116529-60-3
C7H3BrN2O6
291.013
MFCD00458630
BBKJKUFARRJPRP-UHFFFAOYSA-N
2-bromo-3,5-dinitrobenzoic acid, acmc-1bobs, 2-bromo-3,5-dinitrobenzoicacid, benzoic acid,2-bromo-3,5-dinitro
3769937
C1=C(C=C(C(=C1[N+](=O)[O-])Br)C(=O)O)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 213°C to 216°C
Color White
Quantity 1 g
Beilstein 2146483
Sensitivity Light sensitive
Formula Weight 291.01
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-3,5-dinitrobenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'