missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3,4-difluoroaniline, 97%, Thermo Scientific Chemicals

Catalog Number 15438468
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1092349-87-5
C6H4BrF2N
208.01
MFCD11035924
ZGZURPPCPVPDMX-UHFFFAOYSA-N
2-bromo-3,4-difluoro aniline, 2-bromo-3,4-difluorophenylamine, 2-bromo-3,4-difluorobenzenamine
22731604
2-bromo-3,4-difluoroaniline
NC1=C(Br)C(F)=C(F)C=C1
This item is not returnable. View return policy
1092349-87-5
C6H4BrF2N
208.01
MFCD11035924
ZGZURPPCPVPDMX-UHFFFAOYSA-N
2-bromo-3,4-difluoro aniline, 2-bromo-3,4-difluorophenylamine, 2-bromo-3,4-difluorobenzenamine
22731604
2-bromo-3,4-difluoroaniline
NC1=C(Br)C(F)=C(F)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN2811
Formula Weight 208
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Bromo-3,4-difluoroaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'