missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-2-nitropropane-1,3-diol, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
2-Bromo-2-nitropropane-1,3-diol Cosmetics, Preservatives for milk samples, Topical medications and also provide wide spectrum of antibacterial activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-2-nitropropane-1,3-diol Cosmetics, Preservatives for milk samples, Topical medications and also provide wide spectrum of antibacterial activity.
Solubility
Soluble in water(280g/L).
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2-Bromo-2-nitropropane-1,3-diol Cosmetics, Preservatives for milk samples, Topical medications and also provide wide spectrum of antibacterial activity.
Solubility
Soluble in water(280g/L).
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | ∼122°C to 132°C |
pH | 5 to 7 |
Quantity | 100 g |
UN Number | UN3241 |
Beilstein | 1705868 |
Merck Index | 14,1447 |
Solubility Information | Soluble in water(280g/L). |
Formula Weight | 199.99 |
Percent Purity | ≥98% |
Chemical Name or Material | 2-Bromo-2-nitropropane-1,3-diol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'