missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromo-2'-methylacetophenone, 98%

Brand:  Alfa Aesar™ H64651.MD

Code : NEW

Additional Details : CAS Number : 51012-65-8 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15401949

  • £19.89 / 250mg

Estimated Shipment: 27-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications
2-Bromo-2′-methylacetophenone is an aromatic building block for proteomics research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

51012-65-8
213.074
XMGAXELQRATLJP-UHFFFAOYSA-N
12430676
CC1=CC=CC=C1C(=O)CBr
C9H9BrO
MFCD04038955
2-bromo-1-o-tolyl ethanone, 2-bromo-1-2-methylphenyl ethan-1-one, 2'-methylphenacyl bromide, 2-bromo-1-o-tolylethanone, 2-bromo-1-2-methylphenyl ethanone, 2-methylphenacyl bromide, 2-bromo-1-o-tolyl-ethanone, 2-bromo-2'-methylacetophenone, ethanone, 2-bromo-1-methylphenyl, 2-bromoacetyl toluene
2-bromo-1-(2-methylphenyl)ethanone

Specifications

2-Bromo-2'-methylacetophenone
250mg
C9H9BrO
508116
Slightly soluble in water.
CC1=CC=CC=C1C(=O)CBr
213.074
213.07
81°C to 83°C
51012-65-8
MFCD04038955
2-bromo-1-o-tolyl ethanone, 2-bromo-1-2-methylphenyl ethan-1-one, 2'-methylphenacyl bromide, 2-bromo-1-o-tolylethanone, 2-bromo-1-2-methylphenyl ethanone, 2-methylphenacyl bromide, 2-bromo-1-o-tolyl-ethanone, 2-bromo-2'-methylacetophenone, ethanone, 2-bromo-1-methylphenyl, 2-bromoacetyl toluene
XMGAXELQRATLJP-UHFFFAOYSA-N
2-bromo-1-(2-methylphenyl)ethanone
12430676
98%
Videos
SDS Documents
Certificates