missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2'-methoxyacetophenone, 98%, Thermo Scientific Chemicals

Catalog Number 10664955
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
31949-21-0
C9H9BrO2
229.07
MFCD00000196
GKNCPTLOPRDYMH-UHFFFAOYSA-N
2-bromo-1-2-methoxyphenyl ethanone, 2-bromo-2'-methoxyacetophenone, 2-methoxyphenacyl bromide, o-methoxy phenacylbromide, bromomethyl 2-methoxyphenyl ketone, 2-bromo-1-2-methoxyphenyl ethan-1-one, 2-bromo-2'-methoxy acetophenone, ethanone, 2-bromo-1-methoxyphenyl, alpha-bromo-o-methoxyacetophenone, 2-bromoacetylanisole
123440
2-bromo-1-(2-methoxyphenyl)ethanone
COC1=CC=CC=C1C(=O)CBr
This item is not returnable. View return policy
31949-21-0
C9H9BrO2
229.07
MFCD00000196
GKNCPTLOPRDYMH-UHFFFAOYSA-N
2-bromo-1-2-methoxyphenyl ethanone, 2-bromo-2'-methoxyacetophenone, 2-methoxyphenacyl bromide, o-methoxy phenacylbromide, bromomethyl 2-methoxyphenyl ketone, 2-bromo-1-2-methoxyphenyl ethan-1-one, 2-bromo-2'-methoxy acetophenone, ethanone, 2-bromo-1-methoxyphenyl, alpha-bromo-o-methoxyacetophenone, 2-bromoacetylanisole
123440
2-bromo-1-(2-methoxyphenyl)ethanone
COC1=CC=CC=C1C(=O)CBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 43.0°C to 45.0°C
Color White
Boiling Point 130.0°C (1.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4COCH2Br
Quantity 5 g
Formula Weight 229.07
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Bromo-2'-methoxyacetophenone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromo-2'-methoxyacetophenone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'