Learn More
Thermo Scientific Maybridge 2-Bromo-1H-benzimidazole, 10g, 97%
Brand: Thermo Scientific Maybridge CC09010EA
2-Bromo-1H-benzimidazole, 10g, 97%
Code : 31
Additional Details : CAS Number : 54624-57-6 Weight : 0.06000kg
Brand: Thermo Scientific Maybridge CC09010EA
2-Bromo-1H-benzimidazole, 10g, 97%
Code : 31
Additional Details : CAS Number : 54624-57-6 Weight : 0.06000kg