missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromo-1-(cyanomethyl)imidazole-4,5-dicarbonitrile, 97%

Brand:  Alfa Aesar™ H64856.03

Code : NEW

Additional Details : Weight : 0.00100kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15469196

  • £298.92 / 1g


Estimated Shipment date
from Supplier 23-07-2018
Add to basket

Specifications

Chemical Identifiers

2-Bromo-1-(cyanomethyl)imidazole-4,5-dicarbonitrile
236.03
MFCD27975700
UN3439

Specifications

2-Bromo-1-(cyanomethyl)imidazole-4,5-dicarbonitrile
97%
MFCD27975700
Off-white to pale yellow
Solid
236.03
UN3439
1g
Videos
SDS