missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-(5-methyl-1-phenyl-1H-pyrazol-4-yl)-1-ethanone, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber glass bottle |
Quantity | 250 mg |
Formula Weight | 279.14 |
Percent Purity | 95% |
Chemical Name or Material | 2-Bromo-1-(5-methyl-1-phenyl-1H-pyrazol-4-yl)-1-ethanone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'