missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-(5-chloro-3-methylbenzo[b]thiophen-2-yl)ethan-1-one, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
IUPAC Name | 2-bromo-1-(5-chloro-3-methyl-1-benzothiophen-2-yl)ethanone |
Formula Weight | 303.61 |
Percent Purity | 95% |
Chemical Name or Material | 2-Bromo-1-(5-chloro-3-methylbenzo[b]thiophen-2-yl)ethan-1-one |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'