missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-[4-(trifluoromethyl)phenyl]ethan-1-one, Tech., Thermo Scientific™

Catalog Number 10752133
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
383-53-9
C9H6BrF3O
267.05
MFCD00126489
HEMROKPXTCOASZ-UHFFFAOYSA-N
2-bromo-1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethyl phenacyl bromide, 2-bromo-4'-trifluoromethyl acetophenone, 2-bromo-1-4-trifluoromethyl phenyl ethanone, 2-bromo-4'-trifluoromethyl acetophenon, 4-trifluoromethylphenacyl bromide, ethanone, 2-bromo-1-4-trifluoromethyl phenyl, 4'-trifluoromethyl phenacyl bromide, 2-bromo-4'-trifluoromethylacetophenone
321979
2-bromo-1-[4-(trifluoromethyl)phenyl]ethanone
FC(F)(F)C1=CC=C(C=C1)C(=O)CBr
This item is not returnable. View return policy
383-53-9
C9H6BrF3O
267.05
MFCD00126489
HEMROKPXTCOASZ-UHFFFAOYSA-N
2-bromo-1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethyl phenacyl bromide, 2-bromo-4'-trifluoromethyl acetophenone, 2-bromo-1-4-trifluoromethyl phenyl ethanone, 2-bromo-4'-trifluoromethyl acetophenon, 4-trifluoromethylphenacyl bromide, ethanone, 2-bromo-1-4-trifluoromethyl phenyl, 4'-trifluoromethyl phenacyl bromide, 2-bromo-4'-trifluoromethylacetophenone
321979
2-bromo-1-[4-(trifluoromethyl)phenyl]ethanone
FC(F)(F)C1=CC=C(C=C1)C(=O)CBr
Assay Percent Range Tech.
Packaging Amber Glass Bottle
Linear Formula CF3C6H4C(O)CH2Br
Quantity 1 g
Formula Weight 267.04
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Bromo-1-[4-(trifluoromethyl)phenyl]ethan-1-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'