missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-{4-methyl-2-[4-(trifluoromethyl)phenyl]1,3-thiazol-5-yl}-1-ethanone, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Quantity | 5 g |
Formula Weight | 364.18 |
Percent Purity | 97% |
Chemical Name or Material | 2-Bromo-1-{4-methyl-2-[4-(trifluoromethyl)phenyl]-1, 3-thiazol-5-yl}-1-ethanone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'