missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-[4-methyl-2-(2-pyrazinyl)-1,3-thiazol-5-yl]-1-ethanone, ≥90%, Thermo Scientific™

Catalog Number p-2697922
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
423768-43-8
C10H8BrN3OS
298.158
MFCD03407321
PQWKERRFBZJRSD-UHFFFAOYSA-N
2-bromo-1-4-methyl-2-2-pyrazinyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-pyrazin-2-yl thiazol-5-yl ethanone, ethanone,2-bromo-1-4-methyl-2-2-pyrazinyl-5-thiazolyl, 2-bromo-1-4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl ethanone, 5-bromoacetyl-4-methyl-2-pyrazin-2-yl-1,3-thiazole, 2-bromo-1 4-methyl-2-2-pyrazinyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl ethan-1-one
2776507
2-bromo-1-(4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl)ethanone
CC1=C(SC(=N1)C2=NC=CN=C2)C(=O)CBr
This item is not returnable. View return policy
423768-43-8
C10H8BrN3OS
298.158
MFCD03407321
PQWKERRFBZJRSD-UHFFFAOYSA-N
2-bromo-1-4-methyl-2-2-pyrazinyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-pyrazin-2-yl thiazol-5-yl ethanone, ethanone,2-bromo-1-4-methyl-2-2-pyrazinyl-5-thiazolyl, 2-bromo-1-4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl ethanone, 5-bromoacetyl-4-methyl-2-pyrazin-2-yl-1,3-thiazole, 2-bromo-1 4-methyl-2-2-pyrazinyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl ethan-1-one
2776507
2-bromo-1-(4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl)ethanone
CC1=C(SC(=N1)C2=NC=CN=C2)C(=O)CBr
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 298.16
Percent Purity 90%
Chemical Name or Material 2-Bromo-1-[4-methyl-2-(2-pyrazinyl)-1, 3-thiazol-5-yl]-1-ethanone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'