missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(4-isobutylphenyl)propan-1-one, 97%, Thermo Scientific™

Catalog Number p-2697837
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
80336-64-7
C13H17BrO
269.18
MFCD00831700
SVGMFVCPJFHTRE-UHFFFAOYNA-N
2-bromo-1-4-isobutylphenyl propan-1-one, 2-bromo-1-4-2-methylpropyl phenyl propan-1-one, 1-4-isobutylphenyl-2-bromo-1-propanone, 2-bromo-4'-2-methylprop-1-yl propiophenone, 2-bromo-1-4-2-methylpropyl phenyl-1-propanone, 2-bromanyl-1-4-2-methylpropyl phenyl propan-1-one, 1-propanone,2-bromo-1-4-1,1-dimethylethyl phenyl
2735576
2-bromo-1-[4-(2-methylpropyl)phenyl]propan-1-one
CC(C)CC1=CC=C(C=C1)C(=O)C(C)Br
This item is not returnable. View return policy
80336-64-7
C13H17BrO
269.18
MFCD00831700
SVGMFVCPJFHTRE-UHFFFAOYNA-N
2-bromo-1-4-isobutylphenyl propan-1-one, 2-bromo-1-4-2-methylpropyl phenyl propan-1-one, 1-4-isobutylphenyl-2-bromo-1-propanone, 2-bromo-4'-2-methylprop-1-yl propiophenone, 2-bromo-1-4-2-methylpropyl phenyl-1-propanone, 2-bromanyl-1-4-2-methylpropyl phenyl propan-1-one, 1-propanone,2-bromo-1-4-1,1-dimethylethyl phenyl
2735576
2-bromo-1-[4-(2-methylpropyl)phenyl]propan-1-one
CC(C)CC1=CC=C(C=C1)C(=O)C(C)Br
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3CH(Br)C(O)C6H4CH2CH(CH3)2
Quantity 1 g
Formula Weight 269.18
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-(4-isobutylphenyl)propan-1-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'