missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-[4-(difluoromethoxy)phenyl]ethan-1-one, Maybridge

Brand:  Maybridge TL00461DA

UNSPSC : 12350000

Code : 31

Additional Details : CAS Number : 141134-24-9 Weight : 0.05100kg Transport : UN number : 3261 Chem class : 1a Pack group : III

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10180702

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

2-Bromo-1-[4-(difluoromethoxy)phenyl]ethan-1-one
97%
CHF2OC6H4C(O)CH2Br
97%
C9H7BrF2O2
265.05

Specifications

2-Bromo-1-[4-(difluoromethoxy)phenyl]ethan-1-one
Amber Glass Bottle
97%
CHF2OC6H4C(O)CH2Br
1g
141134-24-9
97%
C9H7BrF2O2
265.05
Documentation

Technical Documents