missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-(3-thienyl)-1-ethanone, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 2-bromo-1-3-thienyl-1-ethanone, 2-bromo-1-thiophen-3-yl ethanone, 3-bromoacetylthiophene, 3-bromoacetyl thiophene, 2-bromo-1-thiophen-3-yl ethan-1-one, 2-bromo-1-3-thienyl ethanone, ethanone,2-bromo-1-3-thienyl, ethanone, 2-bromo-1-3-thienyl, 2-bromo-1-thien-3-yl-1-ethanone |
IUPAC Name | 2-bromo-1-(thiophen-3-yl)ethan-1-one |
Formula Weight | 205.07 |
Percent Purity | 97% |
Chemical Name or Material | 2-Bromo-1-(3-thienyl)-1-ethanone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'