missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(3-thienyl)-1-ethanone, 97%, Maybridge™

Brand:  Maybridge CC13105DE

Code : 31

Additional Details : CAS Number : 1468-82-2 Weight : 0.05500kg Transport : UN number : 3261 Chem class : 8 Pack group : III

Packaging Amber Glass Bottle
Quantity 5g

 View more versions of this product

Product Code. 10041694

  • £333.52 / 5g

Estimated Shipment
30-11-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

1468-82-2
205.069
2-bromo-1-3-thienyl-1-ethanone, 2-bromo-1-thiophen-3-yl ethanone, 3-bromoacetylthiophene, 3-bromoacetyl thiophene, 2-bromo-1-thiophen-3-yl ethan-1-one, 2-bromo-1-3-thienyl ethanone, ethanone,2-bromo-1-3-thienyl, ethanone, 2-bromo-1-3-thienyl, 2-bromo-1-thien-3-yl-1-ethanone
2-bromo-1-thiophen-3-ylethanone
C6H5BrOS
TXEJYUFJFSPCHH-UHFFFAOYSA-N
2776379
C1=CSC=C1C(=O)CBr

Specifications

2-Bromo-1-(3-thienyl)-1-ethanone
5g
97%
2-bromo-1-3-thienyl-1-ethanone, 2-bromo-1-thiophen-3-yl ethanone, 3-bromoacetylthiophene, 3-bromoacetyl thiophene, 2-bromo-1-thiophen-3-yl ethan-1-one, 2-bromo-1-3-thienyl ethanone, ethanone,2-bromo-1-3-thienyl, ethanone, 2-bromo-1-3-thienyl, 2-bromo-1-thien-3-yl-1-ethanone
C1=CSC=C1C(=O)CBr
205.069
205.07
Amber Glass Bottle
1468-82-2
C6H5BrOS
TXEJYUFJFSPCHH-UHFFFAOYSA-N
2-bromo-1-thiophen-3-ylethanone
2776379
97%
Videos
SDS Documents