Learn More
Thermo Scientific Maybridge 2-Bromo-1,3-benzothiazole, 5g, 95%
Brand: Thermo Scientific Maybridge CC06010DE
2-Bromo-1,3-benzothiazole, 5g, 95%
Code : 31
Additional Details : CAS Number : 2516-40-7 Weight : 0.05500kg
Brand: Thermo Scientific Maybridge CC06010DE
2-Bromo-1,3-benzothiazole, 5g, 95%
Code : 31
Additional Details : CAS Number : 2516-40-7 Weight : 0.05500kg